APO455830919 Apollo Scientific Ltd

5-Hydroxypyridine-2-carbonitrile

  • MDL number MFCD07368197
  • Cas Number : 86869-14-9
  • Molecular Weight : 120.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-Hydroxypicolinonitrile, 2-Cyano-5-hydroxypyridine, 6-Cyanopyridin-3-ol
Purity 97
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-hydroxypyridine-2-carbonitrile

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-hydroxypyridine-2-carbonitrile
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart