APO455830920 Apollo Scientific Ltd

4-Amino-2-methoxy-5-nitropyridine

  • MDL number MFCD11977410
  • Cas Number : 127356-38-1
  • Molecular Weight : 169.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-methoxy-5-nitro-4-pyridinamine
Inchi Code 1S/C6H7N3O3/c1-12-6-2-4(7)5(3-8-6)9(10)11/h2-3H,1H3,(H2,7,8)
Inchi Key YVXWVLIBEUMASB-UHFFFAOYSA-N

Description

Synonym 2-Methoxy-5-nitropyridin-4-amine
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-amino-2-methoxy-5-nitropyridine

Safety Information

signalword Warning
pcodes P260;P280
MSDS https://store.apolloscientific.co.uk/product/4-amino-2-methoxy-5-nitropyridine
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart