APO455830922 Apollo Scientific Ltd

3-Chloro-4-(dimethylcarbamoyl)benzeneboronic acid

  • MDL number MFCD07363772
  • Cas Number : 850589-47-8
  • Molecular Weight : 227.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 126-132
Synonym 4-Borono-2-chloro-N,N-dimethylbenzamide
Purity 98
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-chloro-4-dimethylcarbamoylbenzeneboronic-acid

Safety Information

signalword Warning
pcodes P280;P302+P352;P304+P340
MSDS https://store.apolloscientific.co.uk/product/3-chloro-4-dimethylcarbamoylbenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart