APO455830924 Apollo Scientific Ltd

Methyl 2-amino-5-ethyl-1,3-thiazole-4-carboxylate

  • MDL number MFCD00728625
  • Cas Number : 28942-54-3
  • Molecular Weight : 186.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 2-amino-5-ethyl-1,3-thiazole-4-carboxylate
Inchi Code 1S/C7H10N2O2S/c1-3-4-5(6(10)11-2)9-7(8)12-4/h3H2,1-2H3,(H2,8,9)
Inchi Key GIAXMSUPTGGVFG-UHFFFAOYSA-N

Description

Synonym 2-Amino-5-ethyl-4-(methoxycarbonyl)-1,3-thiazole
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-2-amino-5-ethyl-13-thiazole-4-carboxylate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-2-amino-5-ethyl-13-thiazole-4-carboxylate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart