APO455830930 Apollo Scientific Ltd

3-Bromo-4-chloronitrobenzene

  • MDL number MFCD00100437
  • Cas Number : 16588-26-4
  • Molecular Weight : 236.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 58
Synonym 2-Bromo-1-chloro-4-nitrobenzene
Purity 98
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-bromo-4-chloronitrobenzene

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromo-4-chloronitrobenzene
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart