APO455830933 Apollo Scientific Ltd

3-Chloro-4-(N-methylcarbamoyl)benzeneboronic acid

  • MDL number MFCD07363766
  • Cas Number : 850589-39-8
  • Molecular Weight : 213.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 164-170
Iupac Name 3-chloro-4-[(methylamino)carbonyl]phenylboronic acid
Inchi Code 1S/C8H9BClNO3/c1-11-8(12)6-3-2-5(9(13)14)4-7(6)10/h2-4,13-14H,1H3,(H,11,12)
Inchi Key UDPDTEGKPHZNES-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 164-170
Product Link https://store.apolloscientific.co.uk/product/3-chloro-4-n-methylcarbamoylbenzeneboronic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-chloro-4-n-methylcarbamoylbenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart