APO455830935 Apollo Scientific Ltd

4-(5-Ethyl-1,3,4-oxadiazol-2-yl)benzaldehyde

  • MDL number MFCD19104824
  • Cas Number : 1309875-09-9
  • Molecular Weight : 202.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(5-ethyl-1,3,4-oxadiazol-2-yl)benzaldehyde
Inchi Code 1S/C11H10N2O2/c1-2-10-12-13-11(15-10)9-5-3-8(7-14)4-6-9/h3-7H,2H2,1H3
Inchi Key GOCQSPCAXCEKNX-UHFFFAOYSA-N

Description

Synonym 2-Ethyl-5-(4-formylphenyl)-1,3,4-oxadiazole
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-5-ethyl-134-oxadiazol-2-ylbenzaldehyde

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-5-ethyl-134-oxadiazol-2-ylbenzaldehyde
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart