APO455830937 Apollo Scientific Ltd

4-[5-(But-1-yl)-1,3,4-oxadiazol-2-yl]benzaldehyde

  • MDL number MFCD18909698
  • Cas Number : 1061672-63-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-(But-1-yl)-5-(4-formylphenyl)-1,3,4-oxadiazole
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=48713

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=48713
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart