APO455830941 Apollo Scientific Ltd

3-Bromophenylacetylene

  • MDL number MFCD03839983
  • Cas Number : 766-81-4
  • Molecular Weight : 181.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-Bromo-3-ethynylbenzene
Storage temp. Store at 2-8
Physical Form Liquid-Clear
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-bromophenylacetylene

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromophenylacetylene
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart