APO455830946 Apollo Scientific Ltd

5-Isopropoxy-1H-indazole-6-boronic acid

  • MDL number MFCD22376779

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym [5-(Propan-2-yloxy)-1H-indazol-6-yl]boronic acid
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/5-isopropoxy-1h-indazole-6-boronic-acid

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/5-isopropoxy-1h-indazole-6-boronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart