APO455830947 Apollo Scientific Ltd

4-Bromo-1-cyclopropyl-1,3-dihydro-2H-benzimidazol-2-one

  • MDL number MFCD22689777
  • Cas Number : 1416714-00-5
  • Molecular Weight : 253.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-bromo-1-cyclopropyl-1H-benzo[d]imidazol-2-ol
Inchi Code 1S/C10H9BrN2O/c11-7-2-1-3-8-9(7)12-10(14)13(8)6-4-5-6/h1-3,6H,4-5H2,(H,12,14)
Inchi Key CCIKBEPPBQBVCI-UHFFFAOYSA-N

Description

Synonym 4-Bromo-1-cyclopropyl-1H-benzo[d]imidazol-2(3H)-one
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-bromo-1-cyclopropyl-13-dihydro-2h-benzimidazol-2-one

Safety Information

signalword Warning
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromo-1-cyclopropyl-13-dihydro-2h-benzimidazol-2-one
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart