APO455830949 Apollo Scientific Ltd

6-Bromo-1-cyclopropyl-1,3-dihydro-2H-benzimidazol-2-one

  • MDL number MFCD22689771
  • Cas Number : 1416714-45-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 6-Bromo-1-cyclopropyl-1H-benzo[d]imidazol-2(3H)-one
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=72955

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=72955
pictograms GHS07
hstatements H302;H312;H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart