APO455830952 Apollo Scientific Ltd

5-Bromo-4-methyl-2-(methylsulphonyl)pyridine

  • MDL number MFCD28397882
  • Cas Number : 1279106-02-3
  • Molecular Weight : 250.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-bromo-4-methyl-2-methylsulphonylpyridine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromo-4-methyl-2-methylsulphonylpyridine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart