APO455830954 Apollo Scientific Ltd

Methyl 2-amino-4-bromobenzoate

  • MDL number MFCD07783014
  • Cas Number : 135484-83-2
  • Molecular Weight : 230.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 77-79
Iupac Name methyl 2-amino-4-bromobenzoate
Inchi Code 1S/C8H8BrNO2/c1-12-8(11)6-3-2-5(9)4-7(6)10/h2-4H,10H2,1H3
Inchi Key MMSODGJNFCCKAZ-UHFFFAOYSA-N

Description

Synonym 5-Bromo-2-(methoxycarbonyl)aniline, Methyl 4-bromoanthranilate
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 77-79
Product Link https://store.apolloscientific.co.uk/product/methyl-2-amino-4-bromobenzoate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-2-amino-4-bromobenzoate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart