APO455830957 Apollo Scientific Ltd

5-(N-Benzylsulphamoyl)-2-methylbenzeneboronic acid

  • MDL number MFCD07363760
  • Cas Number : 871329-73-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 118-122
Synonym 4-(N-Benzylsulphamoyl)-2-boronotoluene, N-Benzyl-3-borono-4-methylbenzenesulphonamide
Purity 97
Storage temp. Ambient temperature
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=30340

Safety Information

signalword Warning
pcodes P261;P280;P271
MSDS http://www.apolloscientific.co.uk/display_item.php?id=30340
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart