APO455830964 Apollo Scientific Ltd

2,6-Dichloro-1,3-benzoxazole

  • MDL number MFCD07368635
  • Cas Number : 3621-82-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 50
Purity 98
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 110
Product Link https://store.apolloscientific.co.uk/product/26-dichloro-13-benzoxazole

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/26-dichloro-13-benzoxazole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart