APO455830967 Apollo Scientific Ltd

3-Formylphenyl acetate

  • MDL number MFCD00016603
  • Cas Number : 34231-78-2
  • Molecular Weight : 164.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-formylphenyl acetate
density 1.17
Inchi Code 1S/C9H8O3/c1-7(11)12-9-4-2-3-8(5-9)6-10/h2-6H,1H3
Inchi Key GVUMZPWBUAGJBP-UHFFFAOYSA-N

Description

Synonym 3-Acetoxybenzaldehyde, Acetic acid 3-formylphenyl ester
Purity 97%
Density 1.17
Flash Point 138/20mm
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-formylphenyl-acetate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-formylphenyl-acetate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart