APO455830971 Apollo Scientific Ltd

2-Amino-6-hydroxypyridine

  • MDL number MFCD00039717
  • Cas Number : 5154-00-7
  • Molecular Weight : 110.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 210-212
Synonym 6-Hydroxypyridin-2-amine, 6-Aminopyridin-2-ol
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-amino-6-hydroxypyridine

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-amino-6-hydroxypyridine
pictograms GHS06
hstatements H301;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart