APO455830980 Apollo Scientific Ltd

2,3-Dichlorobromobenzene

  • MDL number MFCD00000536
  • Cas Number : 56961-77-4
  • Molecular Weight : 225.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 58-61
Iupac Name 1-bromo-2,3-dichlorobenzene
Inchi Code 1S/C6H3BrCl2/c7-4-2-1-3-5(8)6(4)9/h1-3H
Inchi Key HVKCZUVMQPUWSX-UHFFFAOYSA-N

Description

Synonym 1-Bromo-2,3-dichlorobenzene
Flash Point 243/765mm
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 58-61
Product Link https://store.apolloscientific.co.uk/product/23-dichlorobromobenzene

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/23-dichlorobromobenzene
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart