APO455830990 Apollo Scientific Ltd

3-(1H-Imidazol-4-yl)propanoic acid

  • MDL number MFCD00237415
  • Cas Number : 1074-59-5
  • Molecular Weight : 140.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-(2-Carboxyethyl)-1H-imidazole, 3-(1H-Imidazol-4-yl)propionic acid
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-1h-imidazol-4-ylpropanoic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-1h-imidazol-4-ylpropanoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart