APO455830994 Apollo Scientific Ltd

2-Bromo-4-methylaniline hydrochloride

  • MDL number MFCD00142869
  • Cas Number : 13194-71-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Amino-3-bromotoluene hydrochloride
Storage temp. Ambient temperature
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=30010

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=30010
pictograms GHS06
hstatements H301+H331;H311
Availability details for
Quantity
  • Availability
  • Add to Cart