APO455830997 Apollo Scientific Ltd

5-Bromo-2-chlorobenzonitrile

  • MDL number MFCD00672948
  • Cas Number : 57381-44-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 133
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/5-bromo-2-chlorobenzonitrile

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromo-2-chlorobenzonitrile
pictograms GHS06
hstatements H301+H311+H331;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart