APO455830998 Apollo Scientific Ltd

4-Bromo-1-chloro-2-ethylbenzene

  • MDL number MFCD00672954
  • Cas Number : 289039-22-1
  • Molecular Weight : 219.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-bromo-1-chloro-2-ethylbenzene
Inchi Code 1S/C8H8BrCl/c1-2-6-5-7(9)3-4-8(6)10/h3-5H,2H2,1H3
Inchi Key ILQNJGBEXZUYOI-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-bromo-1-chloro-2-ethylbenzene

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromo-1-chloro-2-ethylbenzene
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart