APO455831004 Apollo Scientific Ltd

7-Bromo-5-methyl-1H-indazole

  • MDL number MFCD05664004
  • Cas Number : 885272-97-9
  • Molecular Weight : 211.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 7-bromo-5-methyl-1H-indazole
Inchi Code 1S/C8H7BrN2/c1-5-2-6-4-10-11-8(6)7(9)3-5/h2-4H,1H3,(H,10,11)
Inchi Key NCFBKDULJLGGSG-UHFFFAOYSA-N

Description

Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/7-bromo-5-methyl-1h-indazole

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/7-bromo-5-methyl-1h-indazole
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart