APO455831005 Apollo Scientific Ltd

5-Chloropyrazine-2-carbonitrile

  • MDL number MFCD09607700
  • Cas Number : 36070-75-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 44-45
Synonym 2-Chloro-5-cyanopyrazine, 2-Chloro-5-cyano-1,4-diazine
Purity 97
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Boiling point 242
Product Link http://www.apolloscientific.co.uk/display_item.php?id=48074

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=48074
pictograms GHS07
hstatements H302;H312;H332;H315;H319
Availability details for
Quantity
  • Availability
  • Add to Cart