APO455831007 Apollo Scientific Ltd

7-(Bromomethyl)isoquinoline hydrobromide

  • MDL number MFCD12022338
  • Cas Number : 1203372-02-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 7-(Bromomethyl)-2-azanaphthalene hydrobromide
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=48634

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=48634
pictograms GHS05;GHS07
hstatements H302+H312+H332;H315;H318;H335
Availability details for
Quantity
  • Availability
  • Add to Cart