APO455831010 Apollo Scientific Ltd

(2,3-Dichlorophenyl)methanesulphonyl chloride

  • MDL number MFCD11976102
  • Cas Number : 163295-69-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym alpha-(Chlorosulphonyl)-2,3-dichlorotoluene
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=48632

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=48632
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart