APO455831018 Apollo Scientific Ltd

7-Bromo-2-(hydroxymethyl)benzo[b]thiophene

  • MDL number MFCD12032282
  • Cas Number : 1171926-64-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 77-78
Synonym 7-Bromo-2-(hydroxymethyl)-1-benzothiophene, (7-Bromo-1-benzothiophen-2-yl)methanol
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=49009

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=49009
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart