APO455831024 Apollo Scientific Ltd

Ethyl [5-(morpholin-4-yl)tetrazol-2-yl]acetate

  • MDL number MFCD00052626
  • Cas Number : 175205-05-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-[2-(2-Ethoxy-2-oxoethyl)-2H-tetrazol-5-yl]morpholine
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=30075

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=30075
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart