APO455831027 Apollo Scientific Ltd

5-Bromo-1,7-dimethyl-1H-indazole

  • MDL number MFCD11226595
  • Cas Number : 1092352-34-5
  • Molecular Weight : 225.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-bromo-1,7-dimethyl-1H-indazole
Inchi Code 1S/C9H9BrN2/c1-6-3-8(10)4-7-5-11-12(2)9(6)7/h3-5H,1-2H3
Inchi Key LKDVGRQGTBBLIS-UHFFFAOYSA-N

Description

Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-bromo-17-dimethyl-1h-indazole

Safety Information

signalword Warning
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromo-17-dimethyl-1h-indazole
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart