APO455831034 Apollo Scientific Ltd

4-Bromo-1H-indazole, N1-BOC protected

  • MDL number MFCD11110271
  • Cas Number : 926922-37-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym tert-Butyl 4-bromo-1H-indazole-1-carboxylate
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-bromo-1h-indazole-n1-boc-protected

Safety Information

signalword Warning
pcodes P280;P301+P312
MSDS https://store.apolloscientific.co.uk/product/4-bromo-1h-indazole-n1-boc-protected
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart