APO455831038 Apollo Scientific Ltd

6-Bromo-1-methyl-1H-indazole

  • MDL number MFCD09878569
  • Cas Number : 590417-94-0
  • Molecular Weight : 211.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 48-50
Iupac Name 6-bromo-1-methyl-1H-indazole
Inchi Code 1S/C8H7BrN2/c1-11-8-4-7(9)3-2-6(8)5-10-11/h2-5H,1H3
Inchi Key SVVSOIGNROPKMS-UHFFFAOYSA-N

Description

Purity 98%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 48-50
Product Link https://store.apolloscientific.co.uk/product/6-bromo-1-methyl-1h-indazole

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/6-bromo-1-methyl-1h-indazole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart