APO455831041 Apollo Scientific Ltd

3-(1H-Pyrazol-1-yl)benzeneboronic acid, pinacol ester

  • MDL number MFCD07368520
  • Cas Number : 852227-94-2
  • Molecular Weight : 270.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 75-76
Iupac Name 1-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-1H-pyrazole
Inchi Code 1S/C15H19BN2O2/c1-14(2)15(3,4)20-16(19-14)12-7-5-8-13(11-12)18-10-6-9-17-18/h5-11H,1-4H3
Inchi Key GVSCNAOZDQCWJJ-UHFFFAOYSA-N

Description

Synonym 1-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-1H-pyrazole
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 75-76
Product Link https://store.apolloscientific.co.uk/product/3-1h-pyrazol-1-ylbenzeneboronic-acid-pinacol-ester

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-1h-pyrazol-1-ylbenzeneboronic-acid-pinacol-ester
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart