APO455831042 Apollo Scientific Ltd

1-Methyl-1H-pyrazole-4-boronic acid

  • MDL number MFCD09414709
  • Cas Number : 847818-55-7
  • Molecular Weight : 125.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-methyl-1H-pyrazol-4-ylboronic acid
Inchi Code 1S/C4H7BN2O2/c1-7-3-4(2-6-7)5(8)9/h2-3,8-9H,1H3
Inchi Key RYGOBSYXIIUFOR-UHFFFAOYSA-N

Description

Synonym 4-Borono-1-methyl-1H-pyrazole
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1-methyl-1h-pyrazole-4-boronic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/1-methyl-1h-pyrazole-4-boronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart