APO455831043 Apollo Scientific Ltd

2-(3-Methoxyphenyl)-2-methylpropanoic acid

  • MDL number MFCD09414710
  • Cas Number : 17653-94-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 57-58
Synonym 2-(3-Methoxyphenyl)-2-methylpropionic acid, 3-(2-Carboxyprop-2-yl)anisole
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/2-3-methoxyphenyl-2-methylpropanoic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-3-methoxyphenyl-2-methylpropanoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart