APO455831046 Apollo Scientific Ltd

4-Bromo-2,6-dichloropyridine

  • MDL number MFCD11848646
  • Cas Number : 98027-80-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 95-102
Purity 97
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-bromo-26-dichloropyridine

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromo-26-dichloropyridine
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart