APO455831048 Apollo Scientific Ltd

2-Amino-6-iodophenol

  • MDL number MFCD16999401
  • Cas Number : 99968-81-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Hydroxy-3-iodoaniline
Purity 96
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/2-amino-6-iodophenol

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-amino-6-iodophenol
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart