APO455831049 Apollo Scientific Ltd

(1H-Indazol-4-yl)acetic acid

  • MDL number MFCD13183061
  • Cas Number : 902131-33-3
  • Molecular Weight : 176.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1H-indazol-4-ylacetic acid
Inchi Code 1S/C9H8N2O2/c12-9(13)4-6-2-1-3-8-7(6)5-10-11-8/h1-3,5H,4H2,(H,10,11)(H,12,13)
Inchi Key JKKMMVLTZRIYDW-UHFFFAOYSA-N

Description

Synonym 2-(1H-Indazol-4-yl)ethanoic acid, 4-(Carboxymethyl)-1H-indazole
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1h-indazol-4-ylacetic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/1h-indazol-4-ylacetic-acid
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart