APO455831056 Apollo Scientific Ltd

2'-Methoxy-3'-methylacetophenone

  • MDL number MFCD11044853
  • Cas Number : 6342-75-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-(2-Methoxy-3-methylphenyl)ethan-1-one, 2-Acetyl-6-methylanisole, 3-Acetyl-2-methoxytoluene
Density 1.061
Storage temp. Ambient temperature
Physical Form Liquid-Oil
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 240
Product Link http://www.apolloscientific.co.uk/display_item.php?id=60702

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=60702
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart