APO455831062 Apollo Scientific Ltd

2-(4-Methylpiperazin-1-yl)aniline

  • MDL number MFCD04035359
  • Cas Number : 180605-36-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 100.8-102.3
Synonym 1-(2-Aminophenyl)-4-methylpiperazine
Purity 97
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/2-4-methylpiperazin-1-ylaniline

Safety Information

signalword Danger
pcodes P260;P280;P310
MSDS https://store.apolloscientific.co.uk/product/2-4-methylpiperazin-1-ylaniline
pictograms GHS05;GHS06
hstatements H314;H301+H311;H331
Availability details for
Quantity
  • Availability
  • Add to Cart