APO455831067 Apollo Scientific Ltd

2-Amino-5-bromo-4-(tert-butyl)-1,3-thiazole hydrobromide

  • MDL number MFCD00052240
  • Cas Number : 175136-77-3
  • Molecular Weight : 316.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 235
Synonym 5-Bromo-4-(tert-butyl)-1,3-thiazol-2-amine hydrobromide
Purity 98
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Boiling point 110-112/ 0.3mm
Product Link https://store.apolloscientific.co.uk/product/2-amino-5-bromo-4-tert-butyl-13-thiazole-hydrobromide

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/2-amino-5-bromo-4-tert-butyl-13-thiazole-hydrobromide
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart