APO455831075 Apollo Scientific Ltd

Pyridine-3-sulphonyl chloride hydrochloride

  • MDL number MFCD04035552
  • Cas Number : 42899-76-3
  • Molecular Weight : 214.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 144-145
Iupac Name 3-pyridinesulfonyl chloride hydrochloride
Inchi Code 1S/C5H4ClNO2S.ClH/c6-10(8,9)5-2-1-3-7-4-5;/h1-4H;1H
Inchi Key VIVPWOMJFLICOZ-UHFFFAOYSA-N

Description

Synonym 3-(Chlorosulphonyl)pyridine hydrochloride
Storage temp. Store at -20
Physical Form Solid
Shipping Temperature Shipped with Ice Packs
Country of Origin GB
Boiling point 144-145
Product Link https://store.apolloscientific.co.uk/product/pyridine-3-sulphonyl-chloride-hydrochloride

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/pyridine-3-sulphonyl-chloride-hydrochloride
pictograms GHS05
hstatements H314;EUH014;EUH029
Availability details for
Quantity
  • Availability
  • Add to Cart