APO455831077 Apollo Scientific Ltd

4-(2-Methyl-1,3-thiazol-4-yl)benzenesulphonyl chloride

  • MDL number MFCD07368551
  • Cas Number : 852180-73-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 108-112
Synonym 4-[4-(Chlorosulphonyl)phenyl]-2-methyl-1,3-thiazole
Purity 97
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=30185

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=30185
pictograms GHS05;GHS07
hstatements H302;H314;H335
Availability details for
Quantity
  • Availability
  • Add to Cart