APO455831081 Apollo Scientific Ltd

3-Amino-2-chloro-6-methylphenol

  • MDL number MFCD03094650
  • Cas Number : 84540-50-1
  • Molecular Weight : 157.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 75-77
Synonym 4-Amino-3-chloro-2-hydroxytoluene, 2-Chloro-3-hydroxy-4-methylaniline, 5-Amino-6-chloro-o-cresol
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-amino-2-chloro-6-methylphenol

Safety Information

signalword Warning
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-amino-2-chloro-6-methylphenol
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart