APO455831083 Apollo Scientific Ltd

5-(Bromoacetyl)benzo[b]thiophene

  • MDL number MFCD07368508
  • Cas Number : 1131-87-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 48-50
Synonym 1-(Benzo[b]thiophen-5-yl)-2-bromoethan-1-one, 5-(Bromoacetyl)-1-benzothiophene
Purity 97
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 118-121/20mm
Product Link https://store.apolloscientific.co.uk/product/5-bromoacetylbenzobthiophene

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromoacetylbenzobthiophene
pictograms GHS05;GHS07
hstatements H302;H314
Availability details for
Quantity
  • Availability
  • Add to Cart