APO455831087 Apollo Scientific Ltd

6-(Hydroxymethyl)imidazo[1,2-a]pyridine

  • MDL number MFCD07368521
  • Cas Number : 132213-07-1
  • Molecular Weight : 148.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 104-108
Iupac Name imidazo[1,2-a]pyridin-6-ylmethanol
Inchi Code 1S/C8H8N2O/c11-6-7-1-2-8-9-3-4-10(8)5-7/h1-5,11H,6H2
Inchi Key IEOADZOTWIMSMC-UHFFFAOYSA-N

Description

Synonym Imidazo[1,2-a]pyridin-6-ylmethanol
Purity 97%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 104-108
Product Link https://store.apolloscientific.co.uk/product/6-hydroxymethylimidazo12-apyridine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/6-hydroxymethylimidazo12-apyridine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart