APO455831104 Apollo Scientific Ltd

2-Bromo-4-iodopyridine

  • MDL number MFCD03085768
  • Cas Number : 100523-96-4
  • Molecular Weight : 283.89

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 61
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromo-4-iodopyridine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-4-iodopyridine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart