APO455831107 Apollo Scientific Ltd

4-(Methylsulphonyl)aniline hydrochloride

  • MDL number MFCD00216490
  • Cas Number : 177662-76-9
  • Molecular Weight : 207.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Aminophenyl methyl sulphone hydrochloride
Purity 95
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-methylsulphonylaniline-hydrochloride

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-methylsulphonylaniline-hydrochloride
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart