APO455831131 Apollo Scientific Ltd

3-Bromo-2-iodoanisole

  • MDL number MFCD18089327
  • Cas Number : 450412-22-3
  • Molecular Weight : 312.93

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 61.5-65
Synonym 1-Bromo-2-iodo-3-methoxybenzene, 3-Bromo-2-iodophenyl methyl ether
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-bromo-2-iodoanisole

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromo-2-iodoanisole
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart