APO455831137 Apollo Scientific Ltd

3-Iodophenacyl bromide

  • MDL number MFCD06666937
  • Cas Number : 61858-38-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 62-65
Synonym 2-Bromo-1-(3-iodophenyl)ethan-1-one
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=66004

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=66004
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart